Thermo Scientific Chemicals
Azadibenzocyclooctyne-PEG4-maleimide, Thermo Scientific Chemicals
Catalog number: J65673.MC
100 mg, Each
Thermo Scientific Chemicals
Azadibenzocyclooctyne-PEG4-maleimide, Thermo Scientific Chemicals
Catalog number: J65673.MC
100 mg, Each
Quantity
Catalog number: J65673.MC
also known as J65673-MC
Price (USD)
Quantity
-
Have Questions?
Description
Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG spacer adds hydrophilicity to the product to decrease aggregation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG spacer adds hydrophilicity to the product to decrease aggregation.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.