2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals
2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number J65770.03
also known as J65770-03
Price (USD)/ Each
74.65
Online exclusive
82.40 
Save 7.75 (9%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
82.40 
Save 7.75 (9%)
Add to cart
2'-O-Methylguanosine, 99%, Thermo Scientific Chemicals
Catalog numberJ65770.03
Price (USD)/ Each
74.65
Online exclusive
82.40 
Save 7.75 (9%)
-
Add to cart
Specifications
Chemical Name or Material2'-O-Methylguanosine
CAS2140-71-8
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only