Ascorbate oxidase, from Cucurbita sp.
Ascorbate oxidase, from Cucurbita sp.
Thermo Scientific Chemicals
Ascorbate oxidase, from Cucurbita sp.
Have Questions?
Quantity:
1 kU
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65782.4I
also known as J65782-4I
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.90 
Save 9.25 (15%)
-
Add to cart
Quantity:
1 kU
Request bulk or custom format
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.90 
Save 9.25 (15%)
Add to cart
Ascorbate oxidase, from Cucurbita sp.
Catalog numberJ65782.4I
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.90 
Save 9.25 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Recommended StorageStore at -20°C
EINECS Number232-852-6
Quantity1 kU
FormLyophilized
Unit SizeEach
Ascorbate oxidase, from Cucurbita sp., may be used to study oxidative stress and heat stress response and tolerance. Also used in ascorbic acid assays to study the heat stress response of Arabidopsis and useful for enzymatic determination of ascorbic acid and for eliminating the interference of ascorbic acid in clinical analysis.