N4-Benzoylcytidine, 99%
N4-Benzoylcytidine, 99%
Thermo Scientific Chemicals

N4-Benzoylcytidine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number J65790.06
also known as J65790-06
Price (USD)/ Each
83.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
83.30
Add to cart
N4-Benzoylcytidine, 99%
Catalog numberJ65790.06
Price (USD)/ Each
83.30
-
Add to cart
Chemical Identifiers
CAS13089-48-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.5% min.
Water Content (Karl Fischer Titration)5.0% max.
Loss on Drying1.0% max.
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Ilya A Shestopalov; Surajit Sinha; James K Chen. Light-controlled gene silencing in zebrafish embryos. Nature Chemical Biology, 2007,3650 - 651 .
  2. Wing L. Sung. Synthesis of 4-(1,2,4-triazol-1-yl)-pyrimidin-2(1H)-one ribonucleotide and its application in synthesis of oligoribonucleotides. J. Org. Chem, 1982, 47 (19), 3623-3628.