Thermo Scientific Chemicals

Z-Phe-Ala-fluoromethyl ketone

Catalog number: J65795.LB0
5 mg, Each
Thermo Scientific Chemicals

Z-Phe-Ala-fluoromethyl ketone

Catalog number: J65795.LB0
5 mg, Each
Quantity
Catalog number: J65795.LB0
also known as J65795-#0
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105637-38-5
IUPAC Name
benzyl N-{1-[(4-fluoro-3-oxobutan-2-yl)carbamoyl]-2-phenylethyl}carbamate
Molecular Formula
C21H23FN2O4
InChI Key
ASXVEBPEZMSPHB-UHFFFAOYNA-N
SMILES
CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1)C(=O)CF
Appearance (Color)
White to off-white
Form
Powder
Purity
≥95.0%

Description

Cell permeable negative control for FMK-labelled caspase inhibitors. Also an irreversible inhibitor of cathepsins B, L, and S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text