Thermo Scientific Chemicals

Alcohol dehydrogenase, yeast

Catalog number: J65869.9PL
30 kilounit, Each
Thermo Scientific Chemicals

Alcohol dehydrogenase, yeast

Catalog number: J65869.9PL
30 kilounit, Each
Quantity
Catalog number: J65869.9PL
also known as J65869-9+
Price (USD)
Quantity
-
Have Questions?
Impurity content
NADH oxidase: ≤0.001%
Solution Test
Clear and complete (1 mg/mL in water or dilute buffer)
Form
Powder, freeze-dried
Assay from Supplier's CofA
Specific Activity: ≥290 U/mg protein
Comment
Unit definition: The amount of enzyme which causes the reduction of one micromole of NAD+ per minute at 25°C

Description

Alcohol dehydrogenase catalyzes the reaction: RCH2OH +NAD+ ↔ RCHO + NADH + H+ It facilitates the interconversion between alcohols and aldehydes or ketones with the reduction of nicotinamide adenine dinucleotide (NAD+ to NADH). In biotransformation, alcohol dehydrogenases are often used for the synthesis of enantiomerically pure stereoisomers of chiral alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text