Bethanechol chloride, 98%, Thermo Scientific Chemicals
Bethanechol chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bethanechol chloride, 98%, Thermo Scientific Chemicals

A cholinergic receptor agonist
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65919.06
also known as J65919-06
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
Bethanechol chloride, 98%, Thermo Scientific Chemicals
Catalog numberJ65919.06
Price (USD)/ Each
116.00
-
Add to cart
Specifications
Chemical Name or MaterialBethanechol chloride
Melting Point210°C to 229°C
CAS590-63-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
View more
A cholinergic receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only