Thermo Scientific Chemicals

Histone Acetyltransferase Inhibitor IV, CPTH2

Catalog number: J65939.MA
10 mg, Each
Thermo Scientific Chemicals

Histone Acetyltransferase Inhibitor IV, CPTH2

Catalog number: J65939.MA
10 mg, Each
Quantity
Catalog number: J65939.MA
also known as J65939-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
357649-93-5
Purity
≥95.0%
Appearance (Color)
Brown
Form
Solid

Description

Histone acetylation plays an important role in regulating the chromatin structure and is tightly regulated by two classes of enzyme, histone acetyltransferases (HAT) and histone deacetylases (HDAC). Deregulated HAT and HDAC activity plays a role in the development of a range of cancers. Consequently, inhibitors of these enzymes have potential as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text