Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 7.0, Thermo Scientific Chemicals
This product is animal origin and can only be shipped within the United States
Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 7.0, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 7.0, Thermo Scientific Chemicals

For research use
Have Questions?
Change viewbuttonViewtableView
Quantity:
500g
50g
100g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65969.36
also known as J65969-36
Price (USD)/ Each
-
Quantity:
500g
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
FormulationLyophilized powder
Recommended StorageKeep cold
pH Range7.0
EINECS Number232-936-2
Quantity500g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only