Thermo Scientific Chemicals

Calpain Inhibitor XI

Catalog number: J65991.MCR
1 mg, Each
Thermo Scientific Chemicals

Calpain Inhibitor XI

Catalog number: J65991.MCR
1 mg, Each
Quantity
Catalog number: J65991.MCR
also known as J65991-M^
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
145731-49-3
IUPAC Name
benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Molecular Formula
C26H40N4O6
InChI Key
TZVQRMYLYQNBOA-UHFFFAOYNA-N
SMILES
CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1
Appearance (Color)
Pale yellow
Form
Solid
Purity
≥95%

Description

A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text