Thermo Scientific Chemicals

D-(+)-Trehalose, anhydrous

Catalog number: J66006.06
5 g, Each
Thermo Scientific Chemicals

D-(+)-Trehalose, anhydrous

Catalog number: J66006.06
5 g, Each
Quantity
Catalog number: J66006.06
also known as J66006-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
99-20-7
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
HDTRYLNUVZCQOY-LBYRQFQKNA-N
SMILES
OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Form
White to off white powder
Assay (unspecified)
≥ 98%
Residue on ignition
≤ 0.1%
Water Content (Karl Fischer Titration)
≤ 1.0%
Optical Rotation
Specific rotation: +194 to + 200 (C = 10, H₂O)

Description

Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text