This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66010.ME
also known as J66010-ME
Price (USD)/ Each
227.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
227.00
Add to cart
Diazoxide, 98%
Catalog numberJ66010.ME
Price (USD)/ Each
227.00
-
Add to cart
Specifications
Quantity500 mg
DescriptionWhite to off-white
Recommended StorageKeep cold
Assay98%
CAS364-98-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.