Thermo Scientific Chemicals

D-Ribose-5-phosphate barium salt hexahydrate, 99%

Catalog number: J66017.03
1 g, Each
Thermo Scientific Chemicals

D-Ribose-5-phosphate barium salt hexahydrate, 99%

Catalog number: J66017.03
1 g, Each
Quantity
Catalog number: J66017.03
also known as J66017-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15673-79-7
IUPAC Name
barium(2+) [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Molecular Formula
C5H9BaO8P
InChI Key
VNFDCDZWBYRPMC-QHEAFLAXSA-L
SMILES
[Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
Appearance (Color)
White to yellow
Assay from Suppliers CofA
≥94.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text