Thermo Scientific Chemicals

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide, Thermo Scientific Chemicals

Catalog number: J66024.09
10 g, Each
Thermo Scientific Chemicals

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide, Thermo Scientific Chemicals

Catalog number: J66024.09
10 g, Each
Quantity
Catalog number: J66024.09
also known as J66024-09
Price (USD)
Quantity
-
Have Questions?

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water. Slightly soluble in ethanol

Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text