Spectinomycin sulfate, 93%, Thermo Scientific Chemicals
Spectinomycin sulfate, 93%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Spectinomycin sulfate, 93%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66034.06
also known as J66034-06
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
Add to cart
Spectinomycin sulfate, 93%, Thermo Scientific Chemicals
Catalog numberJ66034.06
Price (USD)/ Each
119.65
Online exclusive
133.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS23312-56-3
IUPAC Name(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
Molecular FormulaC14H26N2O11S
InChI KeyXGBFWQUQYQIFLB-MTTMTQIXSA-N
SMILESOS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Infrared spectrumConforms
Sulfated Ash≤1.0%
FormPowder
Assay from Supplier's CofA≥92.0%
View more
Targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only