Thermo Scientific Chemicals

Tobramycin, 900μg/mg, Thermo Scientific Chemicals

Catalog number: J66040.03
1 g, Each
Thermo Scientific Chemicals

Tobramycin, 900μg/mg, Thermo Scientific Chemicals

Catalog number: J66040.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66040.03
also known as J66040-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32986-56-4
IUPAC Name
(2S,3R,4S,5S,6R)-4-amino-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-6-(hydroxymethyl)oxane-3,5-diol
Molecular Formula
C18H37N5O9
InChI Key
NLVFBUXFDBBNBW-PBSUHMDJSA-N
SMILES
NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](N)C[C@@H]1O
Form
Powder
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥ 900µg/mg
Water Content
≤10.0%

Description

Primarily targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text