Thermo Scientific Chemicals

cis-5,8,11,14,17-Eicosapentaenoic acid

Catalog number: J66060.MC
100 mg, Each
Thermo Scientific Chemicals

cis-5,8,11,14,17-Eicosapentaenoic acid

Catalog number: J66060.MC
100 mg, Each
Quantity
Catalog number: J66060.MC
also known as J66060-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10417-94-4
IUPAC Name
(5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
Molecular Formula
C20H30O2
InChI Key
JAZBEHYOTPTENJ-JLNKQSITSA-N
SMILES
CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O
Appearance (Color)
Colorless
Assay (unspecified)
>98%
Form
Oil

Description

Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text