Thermo Scientific Chemicals
Gibberellic Acid A4, 90%, Thermo Scientific Chemicals
Catalog number: J66065.LB0
5 mg, Each
Thermo Scientific Chemicals
Gibberellic Acid A4, 90%, Thermo Scientific Chemicals
Catalog number: J66065.LB0
5 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66065.LB0
also known as J66065-#0
Price (USD)
Quantity
-
Have Questions?
Description
Used for its stimulating effect on cell division and cell elongation, seed germination and flowering in long day plants
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Used for its stimulating effect on cell division and cell elongation, seed germination and flowering in long day plants