Chemical Name or Material1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose
CAS144490-03-9
Melting Point80°C to 83°C
ColorWhite
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.