Thermo Scientific Chemicals

Poloxamer 188, Thermo Scientific Chemicals

Catalog number: J66087.A1
1 kg, Each
Thermo Scientific Chemicals

Poloxamer 188, Thermo Scientific Chemicals

Catalog number: J66087.A1
1 kg, Each
Quantity
Catalog number: J66087.A1
also known as J66087-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-11-6
IUPAC Name
2-methyloxirane; oxirane
Molecular Formula
C5H10O2
InChI Key
RVGRUAULSDPKGF-UHFFFAOYNA-N
SMILES
C1CO1.CC1CO1
Appearance (Color)
White to cream
Form
Powder or crystalline powder or flakes
Identification (FTIR)
Conforms
Heavy Metals
≤ 20 ppm

Description

Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text