Thermo Scientific Chemicals

4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: J66091.06
5 g, Each
Thermo Scientific Chemicals

4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: J66091.06
5 g, Each
Quantity
Catalog number: J66091.06
also known as J66091-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3368-04-5
IUPAC Name
[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]phosphonic acid
Molecular Formula
C10H9O6P
InChI Key
BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES
CC1=CC(=O)OC2=CC(OP(O)(O)=O)=CC=C12
Water Content (Karl Fischer Titration)
≤1.0%
Form
Powder
Proton NMR
Conforms to structure
Assay from Supplier's CofA
≥99.0% (HPLC)
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text