4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals
4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number J66091.MD
also known as J66091-MD
Price (USD)/ Each
36.30
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
36.30
Add to cart
4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals
Catalog numberJ66091.MD
Price (USD)/ Each
36.30
-
Add to cart
Chemical Identifiers
CAS3368-04-5
IUPAC Name[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]phosphonic acid
Molecular FormulaC10H9O6P
InChI KeyBCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC2=CC(OP(O)(O)=O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.0%
FormPowder
Proton NMRConforms to structure
Assay from Supplier's CofA≥99.0% (HPLC)
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only