Thermo Scientific Chemicals
4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals
Catalog number: J66102.06
5 g, Each
Thermo Scientific Chemicals
4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals
Catalog number: J66102.06
5 g, Each
Quantity
Catalog number: J66102.06
also known as J66102-06
Price (USD)
Quantity
-
Have Questions?
Description
It is used in the treatment of ischemic cardiovascular and cerebrovascular diseases. Also used in Chinese medicine in the treatment of headache and insomnia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.