Exendin (9-39)
Thermo Scientific Chemicals

Exendin (9-39)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66126.EXD
also known as J66126-!D
Price (USD)/ Each
267.65
Online exclusive
297.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
267.65
Online exclusive
297.00 
Save 29.35 (10%)
Add to cart
Exendin (9-39)
Catalog numberJ66126.EXD
Price (USD)/ Each
267.65
Online exclusive
297.00 
Save 29.35 (10%)
-
Add to cart
Specifications
Purification MethodHPLC
FormatSolid
For Use With (Application)Soluble in water at 1mg/ml
Molecular Weight (g/mol)3369.75
Product TypeExendin (9-39)
View more
Exendin-3 (9-39) is a potent and selective GLP-1 receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C