Ribostamycin sulfate, 630^mg/mg, Thermo Scientific Chemicals
Ribostamycin sulfate, 630^mg/mg, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ribostamycin sulfate, 630^mg/mg, Thermo Scientific Chemicals

Used as a broad spectrum antibiotic targeting both Gram-positive and Gram-negative bacteria. Ribostamycin is a broad-spectrum antimicrobial isolated from Streptomyces ribosifidicus. It is used in pharmacokinetic studies.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66155.MD
also known as J66155-MD
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
Add to cart
Ribostamycin sulfate, 630^mg/mg, Thermo Scientific Chemicals
Catalog numberJ66155.MD
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
CAS74195-65-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (unspecified)630?g/mg min.
Loss on Drying5.0% max.
pH6.0 - 8.0
FormPowder
Used in pharmacokinetic studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only