Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Chemical Name or Material | Chondroitin sulfate A sodium salt |
Color | White |
Recommended Storage | Keep cold |
TSCA | No |
MDL Number | MFCD00130792 |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Percent Purity | 90% |
Physical Form | Powder |
Unit Size | Each |
Chemical Name or Material | Chondroitin sulfate A sodium salt |
Color | White |
Recommended Storage | Keep cold |
TSCA | No |
MDL Number | MFCD00130792 |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
Percent Purity | 90% |
Physical Form | Powder |
Unit Size | Each |
Chemical Name or Material | Chondroitin sulfate A sodium salt |
Color | White |
Recommended Storage | Keep cold |
TSCA | No |
MDL Number | MFCD00130792 |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Percent Purity | 90% |
Physical Form | Powder |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
J66156.14 also known as J66156-14 | Each | 25 g | 84.60 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
J66156.06 also known as J66156-06 | Each | 5 g | 31.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
J66156.22 also known as J66156-22 | Each | 100 g | 240.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|