Thermo Scientific Chemicals

4-Methylumbelliferyl sulfate potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J66160.ME
500 mg, Each
Thermo Scientific Chemicals

4-Methylumbelliferyl sulfate potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J66160.ME
500 mg, Each
Quantity
Catalog number: J66160.ME
also known as J66160-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15220-11-8
IUPAC Name
potassium 4-methyl-2-oxo-2H-chromen-7-yl sulfate
Molecular Formula
C10H7KO6S
InChI Key
CSOCSPXOODWGLJ-UHFFFAOYSA-M
SMILES
[K+].CC1=CC(=O)OC2=CC(OS([O-])(=O)=O)=CC=C12
Appearance (Color)
White
Form
Powder
Purity
≥97.5%
Proton NMR
Conforms

Description

4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.

Solubility
Soluble in water at 5mg/ml

Notes
Excitation wavelength, 334 nm; emission wavelength, 370nm (pH 10.4). Reaction product: excitiation wavelength, 360nm; emission wavelength, 449nm
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text