Thermo Scientific Chemicals

Acrylamide, 99.5%, Molecular Biology Grade

Catalog number: J66184.36
500 g, Each
Thermo Scientific Chemicals

Acrylamide, 99.5%, Molecular Biology Grade

Catalog number: J66184.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66184.36
also known as J66184-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-06-1
IUPAC Name
prop-2-enamide
Molecular Formula
C3H5NO
InChI Key
HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES
NC(=O)C=C
Impurity content
Acrylic acid: 0.005% max
Assay (unspecified)
99.5% min.
DNase
None detected
Impurity content
Heavy metals (as lead): 5ppm max.
Melting Point
82-86?C

Description

Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

Solubility
Partly soluble in water.

Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text