Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
Solubility
Partly soluble in water.
Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Charles Beauchamp; Irwin Fridovich. Superoxide dismutase: Improved assays and an assay applicable to acrylamide gels. Analytical Biochemistry., 1971,44(1), 276-287.
Robert M. Zacharius; Tatiana E. Zell; John H. Morrison; John J. Woodlock. Glycoprotein staining following electrophoresis on acrylamide gels. Analytical Biochemistry, 1969,30(1), 148-152.