[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals
[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 mg
Packaging:
Plastic bottle
Catalog number J66191.MCR
also known as J66191-M^
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
1 mg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals
Catalog numberJ66191.MCR
Price (USD)/ Each
161.00
-
Add to cart
Specifications
Chemical Name or Materialalpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt
CAS75921-69-6
Recommended StorageStore at -20°C
RTECS NumberOS1830200
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only