Thermo Scientific Chemicals

Laminarin, Thermo Scientific Chemicals

Catalog number: J66193.MD
250 mg, Each
Thermo Scientific Chemicals

Laminarin, Thermo Scientific Chemicals

Catalog number: J66193.MD
250 mg, Each
Quantity
Catalog number: J66193.MD
also known as J66193-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9008-22-4
IUPAC Name
(3R,4S,5S,6R)-2-{[(2R,3R,4S,5R)-3,5-dihydroxy-2-(hydroxymethyl)-6-{[(3R,4S,5R,6R)-2,3,5-trihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}oxan-4-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C18H32O16
InChI Key
DBTMGCOVALSLOR-VPNXCSTESA-N
SMILES
OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](CO)OC(O[C@@H]3[C@@H](O)C(O)O[C@H](CO)[C@H]3O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Form
Powder
Appearance (Color)
White to off-white
Optical Rotation
-40 to -50? (c=0.9, H₂O)

Description

Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text