Sucrose diacetate hexaisobutyrate
Sucrose diacetate hexaisobutyrate
Thermo Scientific Chemicals

Sucrose diacetate hexaisobutyrate

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
Catalog number J66195.36
also known as J66195-36
Price (USD)/ Each
127.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
127.00
Add to cart
Sucrose diacetate hexaisobutyrate
Catalog numberJ66195.36
Price (USD)/ Each
127.00
-
Add to cart
Specifications
Chemical Name or MaterialSucrose diacetate hexaisobutyrate
CAS27216-37-1
Melting Point-7°C
ColorColorless to Yellow
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only