Thermo Scientific Chemicals

Glucagon-Like Peptide-1 (7-36) amide, human

Catalog number: J66197.EXD
0.5 mg, Each
Thermo Scientific Chemicals

Glucagon-Like Peptide-1 (7-36) amide, human

Catalog number: J66197.EXD
0.5 mg, Each
Quantity
Catalog number: J66197.EXD
also known as J66197-!D
Price (USD)
Quantity
-
Have Questions?
Form
Lyophilized Powder
Appearance (Color)
White
Assay from Suppliers CofA
Peptide Purity: Report Value
Mass Spec
Exhibits correct MW

Description

Glucagon-Like Peptide-1 is used as a Effector in the hormonal control of insulin secretion. The peptide fragment is secreted from the lower small intestine. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text