Agarose, Genetic Technology Grade
Agarose, Genetic Technology Grade
Thermo Scientific Chemicals

Agarose, Genetic Technology Grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number J66208.22
also known as J66208-22
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
Add to cart
Agarose, Genetic Technology Grade
Catalog numberJ66208.22
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Electroendosmosis: 0.09-0.13
Assay (unspecified)Gel Strength (1%): 1200g/cm2 min.
Assay (unspecified)Gelling temperature: 36?C ? 1.5
Water Content (Karl Fischer Titration)10.0% max.
Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
RUO – Research Use Only