Roxithromycin, 97%, Thermo Scientific Chemicals
Roxithromycin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Roxithromycin, 97%, Thermo Scientific Chemicals

Semi-synthetic macrolide antibiotic.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66213.09
also known as J66213-09
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
Add to cart
Roxithromycin, 97%, Thermo Scientific Chemicals
Catalog numberJ66213.09
Price (USD)/ Each
140.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
165.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS80214-83-1
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
Water Content (Karl Fischer Titration)3.0% max.
Used to block bacterial protein synthesis. Clinically, used to treat respiratory tract, urinary and soft tissue infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only