3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals
3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 mg
Catalog number J66223.LB0
also known as J66223-#0
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals
Catalog numberJ66223.LB0
Price (USD)/ Each
217.00
-
Add to cart
Chemical Identifiers
CAS4084-27-9
IUPAC Name(4S,5R)-4,5,6-trihydroxy-2-oxohexanal
Molecular FormulaC6H10O5
InChI KeyZGCHLOWZNKRZSN-NTSWFWBYSA-N
SMILESOC[C@@H](O)[C@@H](O)CC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Proton NMRConforms to structure
Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents
  • Also used for cross-linking studies as it readily reacts with protein amino groups
RUO – Research Use Only