Cefpodoxime, 98%, Thermo Scientific Chemicals
Cefpodoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefpodoxime, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66225.03
also known as J66225-03
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
Add to cart
Cefpodoxime, 98%, Thermo Scientific Chemicals
Catalog numberJ66225.03
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialCefpodoxime
TypeCefpodoxime
Physical FormPowder
CAS80210-62-4
Health Hazard 1H317-H334
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only