Tazobactam sodium, Thermo Scientific Chemicals
Tazobactam sodium, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tazobactam sodium, Thermo Scientific Chemicals

Beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66226.03
also known as J66226-03
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
Add to cart
Tazobactam sodium, Thermo Scientific Chemicals
Catalog numberJ66226.03
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS89785-84-2
IUPAC Namesodium (2S,3S)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC10H11N4NaO5S
InChI KeyRFMIKMMOLPNEDG-LEFOPYBQSA-M
SMILES[Na+].C[C@]1(CN2C=CN=N2)[C@@H](N2C(CC2=O)S1(=O)=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder or crystals
Water Content (Karl Fischer Titration)≤2.5%
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.0%
pH5.0-7.0
Tazobactam sodium is a beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only