Glucagon Like Peptide-1 (7-37)
Glucagon Like Peptide-1 (7-37)
Thermo Scientific Chemicals

Glucagon Like Peptide-1 (7-37)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66232.EXD
also known as J66232-!D
Price (USD)/ Each
197.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
197.00
Add to cart
Glucagon Like Peptide-1 (7-37)
Catalog numberJ66232.EXD
Price (USD)/ Each
197.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatLyophilized Powder
Molecular Weight (g/mol)3355.70
Product TypeGlucagon Like Peptide-1 (7-37)
Quantity0.5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C