Thermo Scientific Chemicals

Sulbactam, 98%, Thermo Scientific Chemicals

Catalog number: J66235.03
1 g, Each
Thermo Scientific Chemicals

Sulbactam, 98%, Thermo Scientific Chemicals

Catalog number: J66235.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66235.03
also known as J66235-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68373-14-8
IUPAC Name
(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Formula
C8H11NO5S
InChI Key
FKENQMMABCRJMK-RITPCOANSA-N
SMILES
CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
Purity
≥97.5%
Form
Powder

Description

Sulbactam is an antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text