Neurotensin, Guinea pig
Neurotensin, Guinea pig
Thermo Scientific Chemicals

Neurotensin, Guinea pig

For cellular and molecular biology applications
Have Questions?
Quantity:
25 mg
Catalog number J66243.MB
also known as J66243-MB
Price (USD)/ Each
461.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
543.00 
Save 81.35 (15%)
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
461.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
543.00 
Save 81.35 (15%)
Add to cart
Neurotensin, Guinea pig
Catalog numberJ66243.MB
Price (USD)/ Each
461.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
543.00 
Save 81.35 (15%)
-
Add to cart
Specifications
SpeciesGuinea Pig
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)1662.91
Product TypeNeurotensin, guinea pig
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C