Galanin, Human
Thermo Scientific Chemicals

Galanin, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66251.EXD
also known as J66251-!D
Price (USD)/ Each
197.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
197.00
Add to cart
Galanin, Human
Catalog numberJ66251.EXD
Price (USD)/ Each
197.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesHuman
Purification MethodHPLC
FormatPowder
Molecular Weight (g/mol)3157.41
Product TypeGalanin, human
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C