Sulfadoxine, 98%, Thermo Scientific Chemicals
Sulfadoxine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfadoxine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66268.22
also known as J66268-22
Price (USD)/ Each
472.00
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
472.00
Add to cart
Sulfadoxine, 98%, Thermo Scientific Chemicals
Catalog numberJ66268.22
Price (USD)/ Each
472.00
-
Add to cart
Specifications
Chemical Name or MaterialSulfadoxine
CAS2447-57-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only