D-Glucosamine sulfate, 98%
This product is animal origin and can only be shipped within the United States
D-Glucosamine sulfate, 98%
Thermo Scientific Chemicals

D-Glucosamine sulfate, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number J66271.14
also known as J66271-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS29031-19-4
IUPAC Name(2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal; sulfuric acid
Molecular FormulaC6H15NO9S
InChI KeyFGNPLIQZJCYWLE-BTVCFUMJSA-N
SMILESOS(O)(=O)=O.N[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
Assay (unspecified)Specific rotation: +52 to +54°
Heavy Metals≤0.001%
Color (APHA)White to pale cream
Loss on Drying≤0.5%
View more
Novel application of glucosamine to prepare medical agent for treating vertigo. Found in chitin, in mucoproteins, and in mucopolysaccharides. Antiarthritic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble at 1mg/ml in 1M HCl

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Carlos C da Camara; Gregory V Dowless. Glucosamine Sulfate for Osteoarthritis. Molecular Cell, 2013,52(3), 340-352.
  2. V Matheu; Mt Gracia Bara; R Pelta; E Vivas; and M Rubio. Immediate-hypersensitivity reaction to glucosamine sulfate. Allergy, 1999,54(6), 643.