Imipenem, mixture with cilastatin, Thermo Scientific Chemicals
Imipenem, mixture with cilastatin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Imipenem, mixture with cilastatin, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66272.LB0
also known as J66272-#0
Price (USD)/ Each
218.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
218.00
Add to cart
Imipenem, mixture with cilastatin, Thermo Scientific Chemicals
Catalog numberJ66272.LB0
Price (USD)/ Each
218.00
-
Add to cart
Specifications
Chemical Name or MaterialImipenem
TypeImipenem
Physical FormPowder
Recommended StorageStore at -20°C
TSCANo
View more
Inhibits the human enzyme dehydropeptidase