Thermo Scientific Chemicals

Amyloid beta (1-40)

Catalog number: J66277.MCR
1 mg, Each
Thermo Scientific Chemicals

Amyloid beta (1-40)

Catalog number: J66277.MCR
1 mg, Each
Quantity
Catalog number: J66277.MCR
also known as J66277-M^
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
White
Form
Lyophilized Powder
Molecular Weight
Conforms
Purity
≥94.0%

Description

Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer’s disease and neurodegeneration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text