Thermo Scientific Chemicals

Calcium L-threonate, 98%, Thermo Scientific Chemicals

Catalog number: J66279.36
500 g, Each
Thermo Scientific Chemicals

Calcium L-threonate, 98%, Thermo Scientific Chemicals

Catalog number: J66279.36
500 g, Each
Quantity
Catalog number: J66279.36
also known as J66279-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70753-61-6
Appearance (Color)
White to off-white
Form
Powder
Assay from Supplier's CofA
≥97.5%

Description

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text