Thermo Scientific Chemicals

D-Galacturonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: J66282.06
5 g, Each
Thermo Scientific Chemicals

D-Galacturonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: J66282.06
5 g, Each
Quantity
Catalog number: J66282.06
also known as J66282-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91510-62-2
SMILES
*
Molecular Weight (g/mol)
0.00
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Water Content
5.0-12.0%
Purity
≥96.0%

Description

D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

Solubility
Soluble in water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text