Thermo Scientific Chemicals
Pefloxacine methanesulfonate dihydrate, 95%
Catalog number: J66285.09
10 g, Each
Thermo Scientific Chemicals
Pefloxacine methanesulfonate dihydrate, 95%
Catalog number: J66285.09
10 g, Each
Quantity
Catalog number: J66285.09
also known as J66285-09
Price (USD)
Quantity
-
Have Questions?
Description
A broad spectrum antibiotic that targets Gram-negative and -positive bacteria, and also some mycoplasma species