Neuromedin B
Thermo Scientific Chemicals

Neuromedin B

 
Have Questions?
Quantity:
1 mg
Catalog number J66286.MCR
also known as J66286-M^
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
Neuromedin B
Catalog numberJ66286.MCR
Price (USD)/ Each
110.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)1132.30
Product TypeNeuromedin B
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C